siRNA Details
Gene Symbol: LINC00272 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034131.1 | 1 | 262 | |||
GenBank mRNA | AF508909.1 | - | 1 | 262 | ||
BC140780.1 | - | 1 | 112 | 9021648 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00272 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034131.1 | 1 | 262 | |||
GenBank mRNA | AF508909.1 | - | 1 | 262 | ||
BC140780.1 | - | 1 | 112 | 9021648 |