siRNA Details
Gene Symbol: LOC107986711 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017012883.1 | XP_016868372.1 | 1 | 107 | ||
XM_017012884.1 | XP_016868373.1 | 1 | 107 |
Gene Symbol: GS1-124K5.4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_134570.1 | 1 | 104 | |||
NR_134571.1 | 1 | 104 | ||||
NR_134572.1 | 1 | 104 | ||||
GenBank mRNA | AY726579.1 | - | 1 | 129 | ||
CT005114.1 | - | 1 | 89 | |||
HY009490.1 | - | 1 | 106 | |||
HY106786.1 | - | 1 | 74 |
