siRNA Details
Gene Symbol: GS1-124K5.4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_134570.1 | 2 | 697 | |||
NR_134571.1 | 2 | 510 | ||||
NR_134572.1 | 2 | 506 | ||||
NR_134573.1 | 2 | 281 | ||||
GenBank mRNA | CN310937.1 | - | 1 | 179 | ||
CT005114.1 | - | 2 | 495 | |||
HY009490.1 | - | 2 | 508 | |||
HY053797.1 | - | 2 | 282 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BU595528.1 | - | 2 | 477 | Chr. 7 - 66493727 - 66495474 [+] on Build GRCh38 |
