siRNA Details
| Gene Symbol: LOC101928614 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135181.1 | 1 | 151 | |||
| GenBank mRNA | BC041439.1 | - | 1 | 151 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BI830188.1 | - | 1 | 151 | Chr. 16 - 86720826 - 86721959 [-] on Build GRCh38 | |

LOC101928614
Genomic Map