siRNA Details
| Gene Symbol: KLHL7-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046220.1 | 3 | 1465 | |||
| GenBank mRNA | AK054880.1 | - | 3 | 1464 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KLHL7-AS1| Gene Symbol: KLHL7-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046220.1 | 3 | 1465 | |||
| GenBank mRNA | AK054880.1 | - | 3 | 1464 | ||