siRNA Details
| Gene Symbol: DEPDC1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110671.1 | 3 | 546 | |||
| GenBank mRNA | BC016787.1 | - | 3 | 550 | ||
| BC020917.1 | - | 3 | 526 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BG614832.1 | - | 3 | 528 | Chr. 1 - 68496696 - 68538164 [+] on Build GRCh38 | |

DEPDC1-AS1
Genomic Map