siRNA Details
| Gene Symbol: LOC101929011 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135069.1 | 1, 2 | 127 | |||
| GenBank mRNA | BX093419.1 | - | 1, 2 | 127 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101929011| Gene Symbol: LOC101929011 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135069.1 | 1, 2 | 127 | |||
| GenBank mRNA | BX093419.1 | - | 1, 2 | 127 | ||