siRNA Details
| Gene Symbol: MRGPRG-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027138.1 | 2 | 394 | |||
| GenBank mRNA | AK093421.1 | - | 2 | 394 | ||
| AK097749.1 | - | 3 | 427 | |||
| BC104971.1 | - | 1 | 99 | |||
| BC104973.1 | - | 1 | 99 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MRGPRG-AS1| Gene Symbol: MRGPRG-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027138.1 | 2 | 394 | |||
| GenBank mRNA | AK093421.1 | - | 2 | 394 | ||
| AK097749.1 | - | 3 | 427 | |||
| BC104971.1 | - | 1 | 99 | |||
| BC104973.1 | - | 1 | 99 | |||