siRNA Details
| Gene Symbol: LINC00382 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120413.1 | 5, 6 | 516 | |||
| GenBank mRNA | BC036310.1 | - | 5, 6 | 518 | ||
| BG717866.1 | - | 5, 6 | 523 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00382| Gene Symbol: LINC00382 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120413.1 | 5, 6 | 516 | |||
| GenBank mRNA | BC036310.1 | - | 5, 6 | 518 | ||
| BG717866.1 | - | 5, 6 | 523 | |||