siRNA Details
| Gene Symbol: C22orf34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017028796.1 | XP_016884285.1 | 7 | 10788 | ||
| GenBank mRNA | AK128192.1 | - | 2 | 1079 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C22orf34| Gene Symbol: C22orf34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017028796.1 | XP_016884285.1 | 7 | 10788 | ||
| GenBank mRNA | AK128192.1 | - | 2 | 1079 | ||