siRNA Details
| Gene Symbol: LINC01227 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104666.1 | 1, 2 | 174 | |||
| GenBank mRNA | BX100000.1 | - | 1, 2 | 174 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01227| Gene Symbol: LINC01227 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104666.1 | 1, 2 | 174 | |||
| GenBank mRNA | BX100000.1 | - | 1, 2 | 174 | ||