siRNA Details
| Gene Symbol: LINC01227 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104665.1 | 3 | 536 | |||
| NR_104666.1 | 2 | 235 | ||||
| GenBank mRNA | AK098656.1 | - | 2 | 932 | ||
| BX100000.1 | - | 2 | 235 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01227| Gene Symbol: LINC01227 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104665.1 | 3 | 536 | |||
| NR_104666.1 | 2 | 235 | ||||
| GenBank mRNA | AK098656.1 | - | 2 | 932 | ||
| BX100000.1 | - | 2 | 235 | |||