siRNA Details
| Gene Symbol: TTC28-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026963.1 | 3, 4 | 432 | |||
| GenBank mRNA | AB051435.1 | - | 2, 3 | 1249 | ||
| AL832597.1 | - | 3, 4 | 387 | |||
| CT841515.1 | - | 790 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TTC28-AS1| Gene Symbol: TTC28-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026963.1 | 3, 4 | 432 | |||
| GenBank mRNA | AB051435.1 | - | 2, 3 | 1249 | ||
| AL832597.1 | - | 3, 4 | 387 | |||
| CT841515.1 | - | 790 | ||||