siRNA Details
| Gene Symbol: LOC100996345 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134922.1 | 2 | 1704 | |||
| NR_134923.1 | 2 | 1681 | ||||
| NR_134924.1 | 2 | 1614 | ||||
| GenBank mRNA | AK057319.1 | - | 2 | 1703 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100996345| Gene Symbol: LOC100996345 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134922.1 | 2 | 1704 | |||
| NR_134923.1 | 2 | 1681 | ||||
| NR_134924.1 | 2 | 1614 | ||||
| GenBank mRNA | AK057319.1 | - | 2 | 1703 | ||