siRNA Details
| Gene Symbol: C2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104191.1 | 3 | 686 | |||
| GenBank mRNA | BX100594.1 | - | 3 | 685 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C2-AS1| Gene Symbol: C2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104191.1 | 3 | 686 | |||
| GenBank mRNA | BX100594.1 | - | 3 | 685 | ||