siRNA Details
Gene Symbol: LINC01483 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109971.1 | 2 | 197 | |||
NR_109972.1 | 2 | 197 | ||||
GenBank mRNA | BC068583.1 | - | 2 | 138 | ||
BC094791.1 | - | 2 | 138 | |||
DA832262.1 | - | 2 | 197 | |||
DA856452.1 | - | 2 | 197 |
