siRNA Details
Gene Symbol: ADIRF-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK128041.1 | - | 3 | 3596 | ||
AL157440.1 | - | 3 | 4373 | |||
BC017676.1 | - | 1 | 1603 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ADIRF-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK128041.1 | - | 3 | 3596 | ||
AL157440.1 | - | 3 | 4373 | |||
BC017676.1 | - | 1 | 1603 |