siRNA Details
| Gene Symbol: LINC00307 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038855.1 | 3 | 369 | |||
| GenBank mRNA | AY063455.1 | - | 3 | 369 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00307| Gene Symbol: LINC00307 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038855.1 | 3 | 369 | |||
| GenBank mRNA | AY063455.1 | - | 3 | 369 | ||