siRNA Details
| Gene Symbol: LOC104613533 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126501.1 | 1 | 66 | |||
| GenBank mRNA | BX089571.1 | - | 1 | 66 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AA399276.1 | - | 1 | 66 | Chr. 15 - 92470243 - 92471546 [-] on Build GRCh38 | |

LOC104613533
Genomic Map