siRNA Details
| Gene Symbol: LOC105373496 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135564.1 | 3 | 410 | |||
| GenBank mRNA | AL557700.3 | - | 3 | 411 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105373496| Gene Symbol: LOC105373496 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135564.1 | 3 | 410 | |||
| GenBank mRNA | AL557700.3 | - | 3 | 411 | ||