siRNA Details
| Gene Symbol: LOC105369891 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135019.1 | 3 | 336 | |||
| GenBank mRNA | BX090745.1 | - | 3 | 336 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105369891| Gene Symbol: LOC105369891 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135019.1 | 3 | 336 | |||
| GenBank mRNA | BX090745.1 | - | 3 | 336 | ||