siRNA Details
| Gene Symbol: LINC00689 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024394.1 | 3 | 2708 | |||
| GenBank mRNA | BC042556.1 | - | 3 | 2642 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00689| Gene Symbol: LINC00689 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024394.1 | 3 | 2708 | |||
| GenBank mRNA | BC042556.1 | - | 3 | 2642 | ||