siRNA Details
| Gene Symbol: B3GALT5-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026542.1 | 2 | 596 | |||
| NR_026543.1 | 2 | 596 | ||||
| GenBank mRNA | AF426266.1 | - | 2 | 215 | ||
| AF426267.1 | - | 2 | 215 | |||
| BC062296.1 | - | 2 | 598 | |||
| BC080530.1 | - | 2 | 595 | |||
| BC119737.2 | - | 2 | 189 | |||
| BC119738.2 | - | 2 | 189 | |||
| BG751912.1 | - | 2 | 595 | |||
| DA218783.1 | - | 2 | 375 | |||

B3GALT5-AS1
Genomic Map