siRNA Details
| Gene Symbol: LINC00559 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_047489.1 | 6 | 3776 | |||
| GenBank mRNA | BC043442.1 | - | 6 | 3778 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00559| Gene Symbol: LINC00559 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_047489.1 | 6 | 3776 | |||
| GenBank mRNA | BC043442.1 | - | 6 | 3778 | ||