siRNA Details
| Gene Symbol: STARD7-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046322.1 | 3 | 1309 | |||
| GenBank mRNA | AK057419.1 | - | 3 | 781 | ||
| AK311432.1 | - | 3 | 1324 | |||
| BC101730.1 | - | 1 | 24 | 8069236 | ||
| BC101732.1 | - | 1 | 24 | 8069238 | ||

STARD7-AS1
Genomic Map