siRNA Details
| Gene Symbol: PSMD6-AS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038286.1 | 1 | 1579 | |||
| GenBank mRNA | AK022323.1 | - | 1 | 314 | ||
| AK023371.1 | - | 1 | 1579 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PSMD6-AS2| Gene Symbol: PSMD6-AS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038286.1 | 1 | 1579 | |||
| GenBank mRNA | AK022323.1 | - | 1 | 314 | ||
| AK023371.1 | - | 1 | 1579 | |||