siRNA Details
Gene Symbol: LOC145694 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_132970.1 | 3 | 1324 | |||
NR_132971.1 | 2 | 1269 | ||||
GenBank mRNA | AK056793.1 | - | 3 | 1324 | ||
CN360125.1 | - | 2 | 443 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC145694 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_132970.1 | 3 | 1324 | |||
NR_132971.1 | 2 | 1269 | ||||
GenBank mRNA | AK056793.1 | - | 3 | 1324 | ||
CN360125.1 | - | 2 | 443 |