siRNA Details
| Gene Symbol: RNF217-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026876.1 | 7 | 3971 | |||
| GenBank mRNA | AL713649.2 | - | 7 | 3973 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RNF217-AS1| Gene Symbol: RNF217-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026876.1 | 7 | 3971 | |||
| GenBank mRNA | AL713649.2 | - | 7 | 3973 | ||