siRNA Details
| Gene Symbol: TSPOAP1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038410.1 | 4 | 666 | |||
| NR_038411.1 | 3 | 461 | ||||
| NR_038412.1 | 3 | 452 | ||||
| NR_038413.1 | 3 | 384 | ||||
| NR_038414.1 | 3 | 634 | ||||
| NR_038415.1 | 2 | 570 | ||||
| NR_038416.1 | 2 | 561 | ||||
| NR_038417.1 | 3 | 433 | ||||
| NR_038418.1 | 3 | 424 | ||||
| GenBank mRNA | AK000271.1 | - | 3 | 528 | ||
| AK090885.1 | - | 4 | 666 | |||
| AK311208.1 | - | 3 | 385 | |||
| AK311306.1 | - | 3 | 423 | |||
| AK311311.1 | - | 3 | 461 | |||
| BC032475.1 | - | 2 | 414 | |||
| BC050403.1 | - | 2 | 414 | |||
| BI820069.1 | - | 3 | 384 | |||

TSPOAP1-AS1
Genomic Map