siRNA Details
| Gene Symbol: SLC8A1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038441.1 | 5 | 244 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BI837006.1 | - | 5 | 242 | Chr. 2 - 39917634 - 40254927 [+] on Build GRCh38 | |

SLC8A1-AS1
Genomic Map