siRNA Details
Gene Symbol: LINC00682 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104143.1 | 2 | 148 | |||
GenBank mRNA | AL521949.3 | - | 2 | 373 | ||
BC025350.1 | - | 2 | 89 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00682 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104143.1 | 2 | 148 | |||
GenBank mRNA | AL521949.3 | - | 2 | 373 | ||
BC025350.1 | - | 2 | 89 |