siRNA Details
| Gene Symbol: LOC102724064 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135761.1 | 2 | 560 | |||
| GenBank mRNA | BM727141.1 | - | 2 | 560 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102724064| Gene Symbol: LOC102724064 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135761.1 | 2 | 560 | |||
| GenBank mRNA | BM727141.1 | - | 2 | 560 | ||