siRNA Details
| Gene Symbol: LINC01570 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110902.1 | 2 | 156 | |||
| GenBank mRNA | DB449025.1 | - | 2 | 78 | ||
| HY009375.1 | - | 2 | 151 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01570| Gene Symbol: LINC01570 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110902.1 | 2 | 156 | |||
| GenBank mRNA | DB449025.1 | - | 2 | 78 | ||
| HY009375.1 | - | 2 | 151 | |||