siRNA Details
| Gene Symbol: LOC101927866 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135560.1 | 3 | 812 | |||
| NR_135561.1 | 2 | 788 | ||||
| GenBank mRNA | BX537734.1 | - | 2 | 748 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927866| Gene Symbol: LOC101927866 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135560.1 | 3 | 812 | |||
| NR_135561.1 | 2 | 788 | ||||
| GenBank mRNA | BX537734.1 | - | 2 | 748 | ||