siRNA Details
| Gene Symbol: LOC105370024 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134996.1 | 2 | 296 | |||
| GenBank mRNA | BM700484.1 | - | 2 | 296 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CR595298.1 | - | 1 | 2132 | Chr. 12 - 119174099 - 119176477 [-] on Build GRCh38 | |

LOC105370024
Genomic Map