siRNA Details
| Gene Symbol: WDR11-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033850.1 | 3 | 826 | |||
| GenBank mRNA | AK094649.1 | - | 3 | 826 | ||
| BC043378.1 | - | 5 | 858 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

WDR11-AS1| Gene Symbol: WDR11-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033850.1 | 3 | 826 | |||
| GenBank mRNA | AK094649.1 | - | 3 | 826 | ||
| BC043378.1 | - | 5 | 858 | |||