siRNA Details
| Gene Symbol: LINC01163 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120619.1 | 2 | 184 | |||
| GenBank mRNA | AK124226.1 | - | 3 | 259 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01163| Gene Symbol: LINC01163 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120619.1 | 2 | 184 | |||
| GenBank mRNA | AK124226.1 | - | 3 | 259 | ||