siRNA Details
| Gene Symbol: LINC01428 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110609.1 | 7 | 659 | |||
| GenBank mRNA | BC038533.1 | - | 7 | 659 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BI830547.1 | - | 658 | Chr. - - [-] on Build | ||

LINC01428
Genomic Map