siRNA Details
| Gene Symbol: LINC01268 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038863.2 | 3 | 2312 | |||
| GenBank mRNA | BC037331.1 | - | 3 | 2317 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01268| Gene Symbol: LINC01268 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038863.2 | 3 | 2312 | |||
| GenBank mRNA | BC037331.1 | - | 3 | 2317 | ||