siRNA Details
| Gene Symbol: LINC01213 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110168.1 | 1 | 32 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01213| Gene Symbol: LINC01213 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110168.1 | 1 | 32 | |||