siRNA Details
| Gene Symbol: LOC101927257 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109965.1 | 4 | 1744 | |||
| GenBank mRNA | AK097769.1 | - | 4 | 1744 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927257| Gene Symbol: LOC101927257 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109965.1 | 4 | 1744 | |||
| GenBank mRNA | AK097769.1 | - | 4 | 1744 | ||