siRNA Details
| Gene Symbol: LOC100507277 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135240.1 | 1 | 112 | |||
| GenBank mRNA | BU852213.1 | - | 1 | 115 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100507277| Gene Symbol: LOC100507277 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135240.1 | 1 | 112 | |||
| GenBank mRNA | BU852213.1 | - | 1 | 115 | ||