siRNA Details
| Gene Symbol: LOC102723722 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017029174.1 | XP_016884663.1 | 4 | 516 | ||
| Gene Symbol: NDUFA6-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034118.2 | 5 | 1293 | |||
| GenBank mRNA | AK123630.1 | - | 5 | 1295 | ||

LOC102723722
Genomic Map