siRNA Details
| Gene Symbol: TTTY22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_001539.1 | 4, 5 | 385 | |||
| GenBank mRNA | AF527834.1 | - | 4, 5 | 385 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AF527834.3 | - | 4, 5 | 385 | Chr. Y - 9801153 - 9813245 [+] on Build GRCh38 | |

TTTY22
Genomic Map