siRNA Details
| Gene Symbol: LOC102723335 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135693.1 | 2 | 188 | |||
| GenBank mRNA | BC032026.1 | - | 2 | 178 | ||
| CR748292.1 | - | 2 | 237 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BG723144.1 | - | 2 | 183 | Chr. 15 - 100558677 - 100559802 [+] on Build GRCh38 | |

LOC102723335
Genomic Map