siRNA Details
| Gene Symbol: MINCR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120682.1 | 1 | 124 | |||
| NR_120683.1 | 1 | 124 | ||||
| GenBank mRNA | AA992445.1 | - | 1 | 112 | ||
| AJ346444.1 | - | 1 | 100 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MINCR| Gene Symbol: MINCR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120682.1 | 1 | 124 | |||
| NR_120683.1 | 1 | 124 | ||||
| GenBank mRNA | AA992445.1 | - | 1 | 112 | ||
| AJ346444.1 | - | 1 | 100 | |||