siRNA Details
| Gene Symbol: LINC00052 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026869.1 | 3 | 210 | |||
| GenBank mRNA | AK056023.1 | - | 3 | 210 | ||
| BC066551.1 | - | 3 | 194 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00052| Gene Symbol: LINC00052 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026869.1 | 3 | 210 | |||
| GenBank mRNA | AK056023.1 | - | 3 | 210 | ||
| BC066551.1 | - | 3 | 194 | |||