siRNA Details
| Gene Symbol: LOC105374313 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_136187.1 | 2 | 213 | |||
| GenBank mRNA | BX091828.1 | - | 2 | 213 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105374313| Gene Symbol: LOC105374313 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_136187.1 | 2 | 213 | |||
| GenBank mRNA | BX091828.1 | - | 2 | 213 | ||