siRNA Details
| Gene Symbol: LOC101927702 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC035195.2 | - | 2 | 712 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927702| Gene Symbol: LOC101927702 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC035195.2 | - | 2 | 712 | ||