siRNA Details
| Gene Symbol: LOC100129029 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135242.1 | 4 | 1671 | |||
| GenBank mRNA | AK126351.1 | - | 4 | 1673 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100129029| Gene Symbol: LOC100129029 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135242.1 | 4 | 1671 | |||
| GenBank mRNA | AK126351.1 | - | 4 | 1673 | ||